Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTTCAGACTTTAAACATCATTA[A/C]ATATGTCACTGCGAGACATCGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LMBRD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LMBRD2 - LMBR1 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007527.1 | 2932 | Missense Mutation | TGT,TTT | C,F 692 | NP_001007528.1 | |
XM_011514162.2 | 2932 | Missense Mutation | TGT,TTT | C,F 692 | XP_011512464.1 | |
XM_017010024.1 | 2932 | Missense Mutation | TGT,TTT | C,F 471 | XP_016865513.1 |