Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGATGGCTAATGCTGCCTGGACC[A/G]CAGGGACTTTTTGTTCGATTTCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611917 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KDM8 - lysine demethylase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NSMCE1 - NSE1 homolog, SMC5-SMC6 complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145080.3 | 900 | Missense Mutation | CGG,TGG | R,W 262 | NP_659547.2 | |
XM_006721023.3 | 900 | Missense Mutation | CGG,TGG | R,W 262 | XP_006721086.1 | |
XM_011545761.2 | 900 | Missense Mutation | CGG,TGG | R,W 299 | XP_011544063.1 | |
XM_011545762.1 | 900 | Missense Mutation | CGG,TGG | R,W 166 | XP_011544064.1 | |
XM_017023040.1 | 900 | Missense Mutation | CGG,TGG | R,W 166 | XP_016878529.1 | |
XM_017023041.1 | 900 | Missense Mutation | CGG,TGG | R,W 166 | XP_016878530.1 | |
XM_017023042.1 | 900 | Missense Mutation | CGG,TGG | R,W 166 | XP_016878531.1 |