Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACACTATAGAAGATGGAGACAG[A/G]GATGGCCACCAAGCCCTGGGGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600547 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KYAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KYAT1 - kynurenine aminotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122671.1 | 1331 | Missense Mutation | CCT,TCT | P,S 380 | NP_001116143.1 | |
NM_001122672.1 | 1331 | Missense Mutation | CCT,TCT | P,S 330 | NP_001116144.1 | |
NM_001287390.1 | 1331 | Missense Mutation | CCT,TCT | P,S 474 | NP_001274319.1 | |
NM_004059.4 | 1331 | Missense Mutation | CCT,TCT | P,S 380 | NP_004050.3 | |
XM_006717311.3 | 1331 | Missense Mutation | CCT,TCT | P,S 414 | XP_006717374.1 | |
XM_011519165.2 | 1331 | Missense Mutation | CCT,TCT | P,S 473 | XP_011517467.1 | |
XM_011519167.2 | 1331 | Missense Mutation | CCT,TCT | P,S 427 | XP_011517469.2 | |
XM_011519168.1 | 1331 | Missense Mutation | CCT,TCT | P,S 380 | XP_011517470.1 | |
XM_011519169.1 | 1331 | Missense Mutation | CCT,TCT | P,S 380 | XP_011517471.1 | |
XM_011519170.2 | 1331 | Missense Mutation | CCT,TCT | P,S 380 | XP_011517472.1 | |
XM_017015260.1 | 1331 | Missense Mutation | CCT,TCT | P,S 487 | XP_016870749.1 | |
XM_017015261.1 | 1331 | Missense Mutation | CCT,TCT | P,S 474 | XP_016870750.1 | |
XM_017015262.1 | 1331 | Missense Mutation | CCT,TCT | P,S 474 | XP_016870751.1 | |
XM_017015263.1 | 1331 | Missense Mutation | CCT,TCT | P,S 473 | XP_016870752.1 | |
XM_017015264.1 | 1331 | Missense Mutation | CCT,TCT | P,S 414 | XP_016870753.1 | |
XM_017015265.1 | 1331 | Missense Mutation | CCT,TCT | P,S 380 | XP_016870754.1 | |
XM_017015266.1 | 1331 | Missense Mutation | CCT,TCT | P,S 380 | XP_016870755.1 | |
XM_017015267.1 | 1331 | Missense Mutation | CCT,TCT | P,S 380 | XP_016870756.1 | |
XM_017015268.1 | 1331 | Missense Mutation | CCT,TCT | P,S 380 | XP_016870757.1 |
SPOUT1 - SPOUT domain containing methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016390.3 | 1331 | Intron | NP_057474.2 | |||
XM_017014804.1 | 1331 | Intron | XP_016870293.1 |