Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGCCTGCTCTTCTTCGAAAACTA[C/T]GAGCACAACACCAGCGTCGTCAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142440 MIM: 600235 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRAMD1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRAMD1A - GRAM domain containing 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HPN - hepsin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCN1B - sodium voltage-gated channel beta subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037.4 | 769 | Silent Mutation | TAC,TAT | Y,Y 132 | NP_001028.1 | |
NM_001321605.1 | 769 | Silent Mutation | TAC,TAT | Y,Y 99 | NP_001308534.1 | |
NM_199037.4 | 769 | Silent Mutation | TAC,TAT | Y,Y 132 | NP_950238.1 |