Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGATGAATGTGAACCAGGTTGCA[A/G]AGAATCTGGCTTTGAGCCACCAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613666 MIM: 608969 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALG11 - ALG11, alpha-1,2-mannosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004127.2 | 755 | UTR 3 | NP_001004127.2 |
LOC107984562 - serine/threonine-protein kinase Nek5-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UTP14C - UTP14, small subunit processome component homolog C (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021645.5 | 755 | Missense Mutation | AAG,GAG | K,E 8 | NP_067677.4 |