Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCTTGCCTCCTCCCCCAGGGAA[G/T]ACCCTGTGGGAGCTGGTGATAGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108730 MIM: 608937 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP2A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP2A1 - ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286075.1 | 325 | Intron | NP_001273004.1 | |||
NM_004320.4 | 325 | Missense Mutation | AAG,AAT | K,N 47 | NP_004311.1 | |
NM_173201.3 | 325 | Missense Mutation | AAG,AAT | K,N 47 | NP_775293.1 |
ATP2A1-AS1 - ATP2A1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH2B1 - SH2B adaptor protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |