Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTTCTTTCTTTCTCAAATCTTG[A/T]GTATTAGCTTTCTTTCTATCAATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601702 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ROCK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ROCK1 - Rho associated coiled-coil containing protein kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005406.2 | 4013 | Silent Mutation | ACA,ACT | T,T 1024 | NP_005397.1 | |
XM_011526137.2 | 4013 | Silent Mutation | ACA,ACT | T,T 561 | XP_011524439.1 |