Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCATGAAATGGAATTTCTTGAC[G/T]CTTTTAACAGAGAAACCCAAGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf163 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf163 - chromosome 6 open reading frame 163 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010868.2 | 1780 | Intron | NP_001010868.2 | |||
XM_005248673.3 | 1780 | Intron | XP_005248730.1 | |||
XM_006715370.3 | 1780 | Intron | XP_006715433.1 | |||
XM_006715372.2 | 1780 | Intron | XP_006715435.1 | |||
XM_011535539.1 | 1780 | Intron | XP_011533841.1 | |||
XM_017010367.1 | 1780 | UTR 5 | XP_016865856.1 |
SMIM8 - small integral membrane protein 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |