Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTTCTGTTTCCACTCGGGGTAC[C/T]TGAGGCACGCCTTGTACATCTTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606646 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARAP1 - ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STARD10 - StAR related lipid transfer domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006645.2 | 1606 | Missense Mutation | AAG,AGG | K,R 221 | NP_006636.2 |