Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGGCTACCTGGGCTAGGGGGCT[A/G]CTGGGCTGAGCAAGACGGGCGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300093 MIM: 300769 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABRE - gamma-aminobutyric acid type A receptor epsilon subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004961.3 | 717 | Nonsense Mutation | CAG,TAG | Q,* 427 | NP_004952.2 | |
XM_011531140.2 | 717 | Nonsense Mutation | CAG,TAG | Q,* 205 | XP_011529442.1 | |
XM_017029386.1 | 717 | Nonsense Mutation | CAG,TAG | Q,* 314 | XP_016884875.1 | |
XM_017029387.1 | 717 | Nonsense Mutation | CAG,TAG | Q,* 282 | XP_016884876.1 | |
XM_017029388.1 | 717 | Nonsense Mutation | CAG,TAG | Q,* 234 | XP_016884877.1 | |
XM_017029389.1 | 717 | UTR 3 | XP_016884878.1 |
MIR224 - microRNA 224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR452 - microRNA 452 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |