Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGACTCATCACAACTTTTCACG[C/G]TGTAAGAGAGGCACAGGAGTGCACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609964 | ||||||||||||||||||||
Literature Links: |
CLEC4D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLEC4D - C-type lectin domain family 4 member D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080387.4 | 337 | Silent Mutation | CGC,CGG | R,R 48 | NP_525126.2 | |
XM_011520632.2 | 337 | Silent Mutation | CGC,CGG | R,R 48 | XP_011518934.1 | |
XM_011520633.2 | 337 | Silent Mutation | CGC,CGG | R,R 48 | XP_011518935.1 |