Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATTTGGGGTTGTCTTGTGCAGT[C/T]GCTTTTGCAAAAACACCAACTGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300169 MIM: 310490 MIM: 300333 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIFM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AIFM1 - apoptosis inducing factor, mitochondria associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130846.3 | 1752 | Silent Mutation | GCA,GCG | A,A 172 | NP_001124318.2 | |
NM_001130847.3 | 1752 | UTR 3 | NP_001124319.1 | |||
NM_004208.3 | 1752 | Silent Mutation | GCA,GCG | A,A 511 | NP_004199.1 | |
NM_145812.2 | 1752 | Silent Mutation | GCA,GCG | A,A 507 | NP_665811.1 |
RAB33A - RAB33A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004794.2 | 1752 | Intron | NP_004785.1 | |||
XM_017029963.1 | 1752 | Intron | XP_016885452.1 |