Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAGAAACTGACTTGAGGCAGTTT[A/C]TTTCTCCATGCCTTAGTTAGATGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300230 MIM: 300165 MIM: 300028 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA5B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CA5B - carbonic anhydrase 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007220.3 | 4792 | UTR 3 | NP_009151.1 | |||
XM_005274442.4 | 4792 | Intron | XP_005274499.1 | |||
XM_017029247.1 | 4792 | Intron | XP_016884736.1 | |||
XM_017029248.1 | 4792 | Intron | XP_016884737.1 | |||
XM_017029249.1 | 4792 | Intron | XP_016884738.1 |
INE2 - inactivation escape 2 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZRSR2 - zinc finger CCCH-type, RNA binding motif and serine/arginine rich 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |