Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTTCCATCCCTCTTCAGGTGGA[G/A]CTACTTATCGATTTTTAGGAGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609477 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMCO4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMCO4 - single-pass membrane protein with coiled-coil domains 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020179.2 | 1530 | Intron | NP_064564.1 | |||
XM_011542907.1 | 1530 | Intron | XP_011541209.1 | |||
XM_011542908.2 | 1530 | Intron | XP_011541210.1 | |||
XM_011542909.1 | 1530 | Intron | XP_011541211.1 | |||
XM_011542910.2 | 1530 | UTR 3 | XP_011541212.1 | |||
XM_011542911.2 | 1530 | UTR 3 | XP_011541213.1 | |||
XM_017018019.1 | 1530 | UTR 3 | XP_016873508.1 | |||
XM_017018020.1 | 1530 | Intron | XP_016873509.1 |