Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTAGGTGGGGTGTATGGGGGAGG[A/C]GCAGGTGATGGCTTGATTCCTCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPNE8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPNE8 - copine 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153634.2 | 1677 | Silent Mutation | NP_705898.1 | |||
XM_011537951.2 | 1677 | Intron | XP_011536253.1 | |||
XM_011537952.2 | 1677 | Intron | XP_011536254.1 | |||
XM_017018851.1 | 1677 | Silent Mutation | XP_016874340.1 | |||
XM_017018852.1 | 1677 | Silent Mutation | XP_016874341.1 |