Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCGCAGATGAAGGTCCCCTGTGG[A/G]ATGGTCTGCAGGGCGCGGACCCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604599 MIM: 606107 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EHMT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EHMT2 - euchromatic histone lysine methyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289413.1 | 3268 | Silent Mutation | ATC,ATT | I,I 1081 | NP_001276342.1 | |
NM_001318833.1 | 3268 | Silent Mutation | ATC,ATT | I,I 856 | NP_001305762.1 | |
NM_006709.4 | 3268 | Silent Mutation | ATC,ATT | I,I 1058 | NP_006700.3 | |
NM_025256.6 | 3268 | Silent Mutation | ATC,ATT | I,I 1024 | NP_079532.5 | |
XM_005248824.3 | 3268 | Silent Mutation | ATC,ATT | I,I 1115 | XP_005248881.1 | |
XM_006714974.1 | 3268 | Silent Mutation | ATC,ATT | I,I 1122 | XP_006715037.1 | |
XM_006714975.1 | 3268 | Silent Mutation | ATC,ATT | I,I 1088 | XP_006715038.1 | |
XM_006714976.1 | 3268 | Silent Mutation | ATC,ATT | I,I 1065 | XP_006715039.1 | |
XM_017010202.1 | 3268 | Silent Mutation | ATC,ATT | I,I 1031 | XP_016865691.1 |
SLC44A4 - solute carrier family 44 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |