Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTACGTGGCCGTGGCATCAGCCAG[A/G]ACGGAGATGAGGCTGGTCTCTGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612202 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102723313 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102723313 - uncharacterized LOC102723313 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOX7 - SRY-box 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031439.3 | 1260 | Silent Mutation | GTC,GTT | V,V 373 | NP_113627.1 |