Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGAAGACCCGCAGCAGGTTTCCA[C/T]GAAGGACACCTTGAAGCTCTTCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609926 MIM: 604119 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPEP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPEP3 - dipeptidase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129758.1 | 1359 | Missense Mutation | CAT,CGT | H,R 422 | NP_001123230.1 | |
NM_022357.3 | 1359 | Missense Mutation | CAT,CGT | H,R 423 | NP_071752.3 |
SLC12A4 - solute carrier family 12 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |