Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGAAGCGAACCACATTCATTTC[A/G]TCCAGGCAAGTCTTGGTGATGTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602320 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NELL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NELL2 - neural EGFL like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145107.1 | 2600 | Silent Mutation | GAC,GAT | D,D 825 | NP_001138579.1 | |
NM_001145108.1 | 2600 | Silent Mutation | GAC,GAT | D,D 775 | NP_001138580.1 | |
NM_001145109.1 | 2600 | Silent Mutation | GAC,GAT | D,D 774 | NP_001138581.1 | |
NM_001145110.1 | 2600 | Silent Mutation | GAC,GAT | D,D 798 | NP_001138582.1 | |
NM_006159.2 | 2600 | Silent Mutation | GAC,GAT | D,D 775 | NP_006150.1 | |
XM_005268905.3 | 2600 | Silent Mutation | GAC,GAT | D,D 775 | XP_005268962.1 | |
XM_011538396.1 | 2600 | Silent Mutation | GAC,GAT | D,D 775 | XP_011536698.1 | |
XM_017019341.1 | 2600 | Silent Mutation | GAC,GAT | D,D 830 | XP_016874830.1 | |
XM_017019342.1 | 2600 | Silent Mutation | GAC,GAT | D,D 780 | XP_016874831.1 | |
XM_017019343.1 | 2600 | Silent Mutation | GAC,GAT | D,D 780 | XP_016874832.1 | |
XM_017019344.1 | 2600 | Silent Mutation | GAC,GAT | D,D 780 | XP_016874833.1 |