Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCGTTCCAGTGACAAAAAGTAT[C/T]CAGTTCTTTTAACATGTACCTTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611326 MIM: 604122 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCPG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCPG1 - cell cycle progression 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204450.1 | 2544 | Missense Mutation | AAT,GAT | N,D 669 | NP_001191379.1 | |
NM_001204451.1 | 2544 | Intron | NP_001191380.1 | |||
NM_004748.4 | 2544 | Missense Mutation | AAT,GAT | N,D 669 | NP_004739.3 | |
NM_020739.3 | 2544 | Missense Mutation | AAT,GAT | N,D 669 | NP_065790.2 |
DYX1C1-CCPG1 - DYX1C1-CCPG1 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGB - phosphatidylinositol glycan anchor biosynthesis class B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |