Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTCTTTCAAGGCGAGGGGCCTCC[A/T]ATCTCAAGCTGTGATACAGGGTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 156490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NME1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NME1 - NME/NM23 nucleoside diphosphate kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000269.2 | 304 | Intron | NP_000260.1 | |||
NM_198175.1 | 304 | Silent Mutation | CCA,CCT | P,P 17 | NP_937818.1 |
NME1-NME2 - NME1-NME2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018136.2 | 304 | Intron | NP_001018146.1 |