Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAGCCTGGTCAGTCTGTAATAT[C/T]GCATGACTCACAGACTCCTTGGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611467 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GBP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GBP5 - guanylate binding protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134486.2 | 1845 | Silent Mutation | GCA,GCG | A,A 472 | NP_001127958.1 | |
NM_052942.3 | 1845 | Silent Mutation | GCA,GCG | A,A 472 | NP_443174.1 |