Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCTGAAACAAGCAGCTTGGGGG[C/T]TTGATTTCATTAGTTACTGGACAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615906 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF219 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF219 - ring finger protein 219 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024546.3 | 1979 | Silent Mutation | AAA,AAG | K,K 640 | NP_078822.3 | |
XM_011535225.1 | 1979 | Silent Mutation | AAA,AAG | K,K 563 | XP_011533527.1 | |
XM_017020743.1 | 1979 | Silent Mutation | AAA,AAG | K,K 563 | XP_016876232.1 |
RNF219-AS1 - RNF219 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |