Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTCAGGAACTCGAGCACGTCGT[C/T]GGGGGATCCCAAGATCACCGGCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611570 MIM: 603027 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBP1 - fructose-bisphosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000507.3 | 1298 | Missense Mutation | AAC,GAC | N,D 323 | NP_000498.2 | |
NM_001127628.1 | 1298 | Missense Mutation | AAC,GAC | N,D 323 | NP_001121100.1 | |
XM_006717005.3 | 1298 | Missense Mutation | AAC,GAC | N,D 241 | XP_006717068.1 |
FBP2 - fructose-bisphosphatase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |