Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGCTGCTGGTGAGCAGCCGCA[C/G]AGGGCAGTGGCGCAGACTTGGAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601408 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KAT6A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KAT6A - lysine acetyltransferase 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305878.1 | 6253 | Intron | NP_001292807.1 | |||
NM_006766.4 | 6253 | Missense Mutation | TCT,TGT | S,C 1866 | NP_006757.2 | |
XM_011544656.2 | 6253 | Missense Mutation | TCT,TGT | S,C 1910 | XP_011542958.1 | |
XM_011544657.2 | 6253 | Missense Mutation | TCT,TGT | S,C 1910 | XP_011542959.1 | |
XM_011544658.2 | 6253 | Missense Mutation | TCT,TGT | S,C 1910 | XP_011542960.1 | |
XM_011544659.2 | 6253 | Missense Mutation | TCT,TGT | S,C 1903 | XP_011542961.1 | |
XM_017013863.1 | 6253 | Missense Mutation | TCT,TGT | S,C 1866 | XP_016869352.1 | |
XM_017013864.1 | 6253 | Missense Mutation | TCT,TGT | S,C 1866 | XP_016869353.1 |