Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCGAACAGGAATCTCCATCTTGG[G/T]TTTCTTTTTGTAAAGTCTGCATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf77 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C12orf77 - chromosome 12 open reading frame 77 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101339.2 | 588 | Missense Mutation | AAA,AAC | K,N 124 | NP_001094809.2 | |
XM_017018966.1 | 588 | Missense Mutation | AAA,AAC | K,N 196 | XP_016874455.1 |
LOC645177 - uncharacterized LOC645177 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |