Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGTTTCTTCTGAATTCTTCTCT[C/T]GCCCAATCTTTCAGGTATTTGCGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610583 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD6 - ankyrin repeat domain 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101929057 - uncharacterized LOC101929057 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LYRM2 - LYR motif containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020466.4 | 186 | Silent Mutation | GCA,GCG | A,A 49 | NP_065199.1 |
MDN1 - midasin AAA ATPase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |