Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATCAGCGCGGACAAGGAGAACA[A/G]CATTAAACTGACCTCGGTGGTGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601974 | ||||||||||||||||||||
Literature Links: |
LOC101928370 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101928370 - uncharacterized LOC101928370 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
S1PR1 - sphingosine-1-phosphate receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320730.1 | 537 | Missense Mutation | AAC,AGC | N,S 44 | NP_001307659.1 | |
NM_001400.4 | 537 | Missense Mutation | AAC,AGC | N,S 44 | NP_001391.2 |