Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCGCCTTGCTCCTCCCCACACTC[C/T]TGGCACAGGGAGAAGGTAAGCAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182139 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HTR3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HTR3A - 5-hydroxytryptamine receptor 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000869.5 | 303 | Silent Mutation | CTG,TTG | L,L 24 | NP_000860.2 | |
NM_001161772.2 | 303 | Intron | NP_001155244.1 | |||
NM_213621.3 | 303 | Silent Mutation | CTG,TTG | L,L 24 | NP_998786.2 |