Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATTCTTTCTTTTCCACTTGTTG[C/T]GTAAGGCCTCCAAACCCAATACCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600824 MIM: 612815 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSRP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSRP3 - cysteine and glycine rich protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003476.4 | 800 | Silent Mutation | ACA,ACG | T,T 187 | NP_003467.1 |
ZDHHC13 - zinc finger DHHC-type containing 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |