Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGCAAGCATCAAGTCTAGCTGT[C/T]AAGTTTTCACCAGCCTACCACCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605380 MIM: 610775 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGF23 - fibroblast growth factor 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020638.2 | 1524 | UTR 3 | NP_065689.1 |
TIGAR - TP53 induced glycolysis regulatory phosphatase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |