Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCCCAGCCCTGGCCTCATCACT[A/G]TGCGGCTCAAATTCCTCAATGATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB16-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASB16-AS1 - ASB16 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ATXN7L3 - ataxin 7 like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101926967 - uncharacterized LOC101926967 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMUB2 - transmembrane and ubiquitin like domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076674.1 | 1116 | Missense Mutation | ATG,GTG | M,V 176 | NP_001070142.1 | |
NM_024107.2 | 1116 | Missense Mutation | ATG,GTG | M,V 156 | NP_077012.2 | |
NM_177441.2 | 1116 | Missense Mutation | ATG,GTG | M,V 156 | NP_803190.2 | |
XM_011525198.2 | 1116 | Missense Mutation | ATG,GTG | M,V 176 | XP_011523500.1 | |
XM_011525200.2 | 1116 | Missense Mutation | ATG,GTG | M,V 229 | XP_011523502.1 | |
XM_011525201.2 | 1116 | Missense Mutation | ATG,GTG | M,V 176 | XP_011523503.1 | |
XM_011525203.2 | 1116 | Missense Mutation | ATG,GTG | M,V 167 | XP_011523505.1 | |
XM_011525204.2 | 1116 | Missense Mutation | ATG,GTG | M,V 167 | XP_011523506.1 | |
XM_011525205.2 | 1116 | Missense Mutation | ATG,GTG | M,V 156 | XP_011523507.1 | |
XM_011525206.2 | 1116 | Missense Mutation | ATG,GTG | M,V 156 | XP_011523508.1 | |
XM_011525207.2 | 1116 | Missense Mutation | ATG,GTG | M,V 156 | XP_011523509.1 | |
XM_011525208.2 | 1116 | Missense Mutation | ATG,GTG | M,V 156 | XP_011523510.1 | |
XM_011525209.2 | 1116 | Missense Mutation | ATG,GTG | M,V 156 | XP_011523511.1 | |
XM_011525210.2 | 1116 | Missense Mutation | ATG,GTG | M,V 119 | XP_011523512.1 | |
XM_011525211.2 | 1116 | Intron | XP_011523513.1 | |||
XM_017025047.1 | 1116 | Missense Mutation | ATG,GTG | M,V 167 | XP_016880536.1 | |
XM_017025048.1 | 1116 | Missense Mutation | ATG,GTG | M,V 156 | XP_016880537.1 | |
XM_017025049.1 | 1116 | Missense Mutation | ATG,GTG | M,V 156 | XP_016880538.1 | |
XM_017025050.1 | 1116 | Missense Mutation | ATG,GTG | M,V 156 | XP_016880539.1 | |
XM_017025051.1 | 1116 | Missense Mutation | ATG,GTG | M,V 156 | XP_016880540.1 | |
XM_017025052.1 | 1116 | Missense Mutation | ATG,GTG | M,V 130 | XP_016880541.1 | |
XM_017025053.1 | 1116 | Missense Mutation | ATG,GTG | M,V 130 | XP_016880542.1 | |
XM_017025054.1 | 1116 | Missense Mutation | ATG,GTG | M,V 119 | XP_016880543.1 | |
XM_017025055.1 | 1116 | Missense Mutation | ATG,GTG | M,V 119 | XP_016880544.1 | |
XM_017025056.1 | 1116 | Intron | XP_016880545.1 |