Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGCTCTCCGTGGAAATCTTCTC[A/G]ATAGATTCCACCACTTCCACTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603307 MIM: 162151 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BFSP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BFSP1 - beaded filament structural protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161705.1 | 1924 | Silent Mutation | ATC,ATT | I,I 503 | NP_001155177.1 | |
NM_001195.4 | 1924 | Silent Mutation | ATC,ATT | I,I 628 | NP_001186.1 | |
NM_001278606.1 | 1924 | Silent Mutation | ATC,ATT | I,I 489 | NP_001265535.1 | |
NM_001278607.1 | 1924 | Silent Mutation | ATC,ATT | I,I 517 | NP_001265536.1 | |
NM_001278608.1 | 1924 | Silent Mutation | ATC,ATT | I,I 489 | NP_001265537.1 | |
XM_017028005.1 | 1924 | Silent Mutation | ATC,ATT | I,I 592 | XP_016883494.1 |
PCSK2 - proprotein convertase subtilisin/kexin type 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |