Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAACTACCCAGCTTTCATGATG[A/G]AAATGAAGCATGTCTTTGAAGACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609810 MIM: 604149 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEG10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PEG10 - paternally expressed 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040152.1 | 936 | Missense Mutation | AAA,GAA | K,E 153 | NP_001035242.1 | |
NM_001172437.2 | 936 | Missense Mutation | AAA,GAA | K,E 229 | NP_001165908.1 | |
NM_001172438.2 | 936 | Missense Mutation | AAA,GAA | K,E 229 | NP_001165909.1 | |
NM_001184961.1 | 936 | Missense Mutation | AAA,GAA | K,E 187 | NP_001171890.1 | |
NM_001184962.1 | 936 | Missense Mutation | AAA,GAA | K,E 187 | NP_001171891.1 | |
NM_015068.3 | 936 | Missense Mutation | AAA,GAA | K,E 153 | NP_055883.2 |
SGCE - sarcoglycan epsilon | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |