Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCCTTGGCCTGGCCTGGCCCAA[C/G]CCCACAGAGACAACAAATCCACCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607627 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BPIFA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BPIFA3 - BPI fold containing family A member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042439.2 | 321 | Missense Mutation | CCC,GCC | P,A 32 | NP_001035904.1 | |
NM_178466.4 | 321 | Missense Mutation | CCC,GCC | P,A 32 | NP_848561.2 |
BPIFA4P - BPI fold containing family A member 4, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |