Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCTATGAGCAGTTCAACTTCAC[C/T]ATGGAGGATGGCCTGGTAAGAAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108960 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR8083 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR8083 - microRNA 8083 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPR1 - natriuretic peptide receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000906.3 | 1512 | Silent Mutation | ACC,ACT | T,T 340 | NP_000897.3 | |
XM_005245218.1 | 1512 | Silent Mutation | ACC,ACT | T,T 340 | XP_005245275.1 | |
XM_017001374.1 | 1512 | Silent Mutation | ACC,ACT | T,T 340 | XP_016856863.1 |