Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGAAACCCTCGAAGGCTCAAAAG[C/T]TGGTCTCTTATCAAGAATACCTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603429 MIM: 143024 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCF1 - ATP binding cassette subfamily F member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GNL1 - G protein nucleolar 1 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRR3 - proline rich 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077497.2 | 978 | Silent Mutation | AGC,AGT | S,S 114 | NP_001070965.1 | |
NM_025263.3 | 978 | Silent Mutation | AGC,AGT | S,S 135 | NP_079539.2 |