Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAATAAAACAACAGAAATTCCT[C/T]TAATTGGGTATGTAATCTTCATAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP47 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP47 - cilia and flagella associated protein 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304548.1 | 453 | Silent Mutation | CTA,TTA | L,L 132 | NP_001291477.1 | |
NM_152632.3 | 453 | Silent Mutation | CTA,TTA | L,L 132 | NP_689845.2 | |
XM_017029452.1 | 453 | Silent Mutation | CTA,TTA | L,L 132 | XP_016884941.1 | |
XM_017029453.1 | 453 | Silent Mutation | CTA,TTA | L,L 132 | XP_016884942.1 |