Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACATGCTTTTGAAAGTACTGAGC[A/G]TGGCTGAAGGCTTTCCCACATTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194551 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF77 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF77 - zinc finger protein 77 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021217.2 | 1581 | Silent Mutation | CAC,CAT | H,H 475 | NP_067040.1 | |
XM_017027081.1 | 1581 | Silent Mutation | CAC,CAT | H,H 295 | XP_016882570.1 | |
XM_017027082.1 | 1581 | Silent Mutation | CAC,CAT | H,H 295 | XP_016882571.1 |