Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGAAATGTCATCAGTGCAGTCACA[A/G]CAGGAGCAGTTGTCCCAGTCAGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615003 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDHD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDHD2 - DDHD domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164232.1 | 546 | Silent Mutation | CAA,CAG | Q,Q 7 | NP_001157704.1 | |
NM_001164234.1 | 546 | Silent Mutation | CAA,CAG | Q,Q 7 | NP_001157706.1 | |
NM_015214.2 | 546 | Silent Mutation | CAA,CAG | Q,Q 7 | NP_056029.2 | |
XM_005273454.2 | 546 | Silent Mutation | CAA,CAG | Q,Q 7 | XP_005273511.1 | |
XM_005273455.3 | 546 | Silent Mutation | CAA,CAG | Q,Q 7 | XP_005273512.1 | |
XM_005273456.3 | 546 | Silent Mutation | CAA,CAG | Q,Q 7 | XP_005273513.1 | |
XM_011544456.1 | 546 | Silent Mutation | CAA,CAG | Q,Q 7 | XP_011542758.1 | |
XM_017013252.1 | 546 | Silent Mutation | CAA,CAG | Q,Q 7 | XP_016868741.1 | |
XM_017013253.1 | 546 | Silent Mutation | CAA,CAG | Q,Q 7 | XP_016868742.1 | |
XM_017013254.1 | 546 | UTR 5 | XP_016868743.1 | |||
XM_017013255.1 | 546 | UTR 5 | XP_016868744.1 | |||
XM_017013256.1 | 546 | UTR 5 | XP_016868745.1 |