Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCCCTTTAATGTTAAGAATGCT[A/G]ATCTCACCGAAGTAGCTGCCATCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123825 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNGA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNGA1 - cyclic nucleotide gated channel alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000087.3 | 1962 | Silent Mutation | ATC,ATT | I,I 547 | NP_000078.2 | |
NM_001142564.1 | 1962 | Silent Mutation | ATC,ATT | I,I 616 | NP_001136036.1 | |
XM_005248049.4 | 1962 | Silent Mutation | ATC,ATT | I,I 622 | XP_005248106.2 | |
XM_011513623.2 | 1962 | Silent Mutation | ATC,ATT | I,I 547 | XP_011511925.1 | |
XM_017007712.1 | 1962 | Silent Mutation | ATC,ATT | I,I 547 | XP_016863201.1 |
LOC101927157 - uncharacterized LOC101927157 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |