Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGGCATTGCATTTTGATTATCT[A/T]CTTTTTTGAGACCCCGGAATGCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607666 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANGPTL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANGPTL5 - angiopoietin like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178127.4 | 1045 | Missense Mutation | GAA,GTA | E,V 292 | NP_835228.2 | |
XM_011542735.2 | 1045 | Missense Mutation | GAA,GTA | E,V 227 | XP_011541037.1 | |
XM_017017466.1 | 1045 | Missense Mutation | GAA,GTA | E,V 152 | XP_016872955.1 |