Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTATCCTTATGGGGGAGCTCTCT[A/G]ATCACCATGACCATCATGATGAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300361 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BEX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BEX3 - brain expressed X-linked 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282674.1 | 647 | Missense Mutation | AAT,GAT | N,D 179 | NP_001269603.1 | |
NM_014380.2 | 647 | Missense Mutation | AAT,GAT | N,D 99 | NP_055195.1 | |
NM_206915.2 | 647 | Missense Mutation | AAT,GAT | N,D 99 | NP_996798.1 | |
NM_206917.2 | 647 | Missense Mutation | AAT,GAT | N,D 89 | NP_996800.1 |