Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAACTGGGAGGAGTTTGACGAACA[C/G]AGAGACTCCCGGAGGAGCATCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8orf74 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C8orf74 - chromosome 8 open reading frame 74 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040032.1 | 238 | Missense Mutation | CAC,CAG | H,Q 36 | NP_001035121.1 | |
XM_011543816.2 | 238 | Missense Mutation | CAC,CAG | H,Q 43 | XP_011542118.1 |
MIR4286 - microRNA 4286 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |