Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCATGAAGTTTTGTTACTGACCT[C/T]TCTTGGACTTTAGCTCTCCATCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154870 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MGP - matrix Gla protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000900.3 | 371 | Silent Mutation | GAA,GAG | E,E 56 | NP_000891.2 | |
NM_001190839.1 | 371 | Silent Mutation | GAA,GAG | E,E 81 | NP_001177768.1 |