Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTATATCCTCATTTTTTAGGCTCA[G/A]GGATACATACACTAACACTGTAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600687 | ||||||||||||||||||||
Literature Links: |
TIAM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TIAM1 - T-cell lymphoma invasion and metastasis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003253.2 | 6077 | UTR 3 | NP_003244.2 | |||
XM_005261037.1 | 6077 | UTR 3 | XP_005261094.1 | |||
XM_005261038.2 | 6077 | UTR 3 | XP_005261095.1 | |||
XM_005261040.2 | 6077 | UTR 3 | XP_005261097.1 | |||
XM_011529711.1 | 6077 | UTR 3 | XP_011528013.1 | |||
XM_011529712.1 | 6077 | UTR 3 | XP_011528014.1 | |||
XM_011529713.1 | 6077 | UTR 3 | XP_011528015.1 | |||
XM_017028447.1 | 6077 | UTR 3 | XP_016883936.1 | |||
XM_017028448.1 | 6077 | UTR 3 | XP_016883937.1 | |||
XM_017028449.1 | 6077 | UTR 3 | XP_016883938.1 | |||
XM_017028450.1 | 6077 | UTR 3 | XP_016883939.1 | |||
XM_017028451.1 | 6077 | UTR 3 | XP_016883940.1 | |||
XM_017028452.1 | 6077 | UTR 3 | XP_016883941.1 | |||
XM_017028453.1 | 6077 | UTR 3 | XP_016883942.1 |