Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGATTGTTAGTGGAACTTCTAC[C/G]CGACACTTACATGCCATGGCCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608259 MIM: 614624 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGF2BP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGF2BP3 - insulin like growth factor 2 mRNA binding protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MALSU1 - mitochondrial assembly of ribosomal large subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138446.1 | 428 | Silent Mutation | ACC,ACG | T,T 132 | NP_612455.1 | |
XM_017011731.1 | 428 | Silent Mutation | ACC,ACG | T,T 37 | XP_016867220.1 |