Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCGCCTCGTGGTTAGTTTGGGCA[A/C]TGTTGATGTACTGAAGCAGCATATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP20A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP20A1 - cytochrome P450 family 20 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177538.2 | 870 | Missense Mutation | AAT,ACT | N,T 83 | NP_803882.1 | |
XM_005246698.3 | 870 | Intron | XP_005246755.1 | |||
XM_005246699.3 | 870 | Intron | XP_005246756.1 | |||
XM_005246701.3 | 870 | Intron | XP_005246758.1 | |||
XM_006712644.3 | 870 | Missense Mutation | AAT,ACT | N,T 65 | XP_006712707.1 | |
XM_006712645.3 | 870 | Missense Mutation | AAT,ACT | N,T 83 | XP_006712708.1 | |
XM_017004555.1 | 870 | Intron | XP_016860044.1 |