Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606073 MIM: 609826 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYSRT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYSRT1 - cysteine rich tail 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199001.3 | 722 | Missense Mutation | ACC,TCC | T,S 79 | NP_945352.3 |
NDOR1 - NADPH dependent diflavin oxidoreductase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF208 - ring finger protein 208 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF224 - ring finger protein 224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC34A3 - solute carrier family 34 member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |