Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCTTGGTCCTAAGAGATTTCGA[C/T]GGGCTTGTTCAAGTTATCATTCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611503 MIM: 610956 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CENPL - centromere protein L | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DARS2 - aspartyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018122.4 | 832 | Silent Mutation | GAC,GAT | D,D 87 | NP_060592.2 | |
XM_006711427.3 | 832 | Silent Mutation | GAC,GAT | D,D 87 | XP_006711490.1 | |
XM_011509711.2 | 832 | Silent Mutation | GAC,GAT | D,D 87 | XP_011508013.1 |